Gene Symbol: Stau1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109907.1 | NP_001103377.1 | 4 | 571 | ||
NM_053436.2 | NP_445888.1 | 4 | 571 | |||
GenBank mRNA | AF227200.1 | - | 4 | 558 | ||
AF290989.1 | - | 4 | 558 | |||
AJ010200.1 | - | 2 | 252 | |||
BC081755.1 | - | 4 | 529 | |||
BC101858.1 | - | 4 | 567 | 7937343 | ||
DN932990.1 | - | 4 | 600 |