Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053498.1 | NP_445950.1 | 10 | 1063 | ||
XM_006235218.2 | XP_006235280.1 | 10 | 1158 | |||
GenBank mRNA | AJ277748.1 | - | 10 | 1063 |