Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053546.1 | NP_445998.1 | 7 | 1460 | ||
XM_006241609.2 | XP_006241671.1 | 7 | 1805 | |||
XM_008765463.1 | XP_008763685.1 | 7 | 989 | |||
XM_008765464.1 | XP_008763686.1 | 7 | 3453 | |||
GenBank mRNA | AB080023.1 | - | 7 | 1460 |