Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001057.2 | NP_001048.2 | 1 | 791 | ||
GenBank mRNA | AK291996.1 | - | 1 | 791 | ||
AK300649.1 | - | 1 | 685 | |||
AY322545.1 | - | 1 | 196 | |||
BC096842.1 | - | 1 | 202 | 6971906 | ||
M57414.1 | - | 1 | 196 |
Set Membership: |
G-protein coupled receptor |