Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001057.2 | NP_001048.2 | 3 | 1235 | ||
GenBank mRNA | AK291996.1 | - | 3 | 1235 | ||
AK300649.1 | - | 3 | 1033 | |||
AY322545.1 | - | 3 | 640 | |||
BC096842.1 | - | 3 | 646 | 6971906 | ||
FJ200479.1 | - | 4 | 532 | |||
M57414.1 | - | 3 | 640 |
Set Membership: |
![]() |