Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbtps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053569.1 | NP_446021.1 | 3 | 600 | ||
XM_006255723.2 | XP_006255785.1 | 2 | 1832 | |||
XM_008772651.1 | XP_008770873.1 | 2 | 1832 | |||
GenBank mRNA | AF094821.1 | - | 3 | 600 | ||
BC061855.1 | - | 3 | 654 |