Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053586.1 | NP_446038.1 | 3 | 263 | ||
GenBank mRNA | BC083179.1 | - | 3 | 276 | 7300877 | |
D10952.1 | - | 3 | 263 | |||
FQ210284.1 | - | 3 | 260 | |||
FQ214227.1 | - | 3 | 266 | |||
FQ215884.1 | - | 3 | 266 | |||
FQ217498.1 | - | 3 | 266 | |||
FQ217781.1 | - | 3 | 284 | |||
FQ217835.1 | - | 3 | 266 | |||
FQ218282.1 | - | 3 | 264 | |||
FQ224825.1 | - | 3 | 277 | |||
X14208.1 | - | 3 | 211 |