Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001058.3 | NP_001049.1 | 4 | 1414 | ||
NM_015727.2 | NP_056542.1 | 4 | 1414 | |||
GenBank mRNA | AK289765.1 | - | 4 | 1383 | ||
AY462098.1 | - | 4 | 829 | |||
BC074911.2 | - | 4 | 851 | 30915523 | ||
BC074912.2 | - | 4 | 851 | 30915310 | ||
BC095528.1 | - | 4 | 826 | |||
M74290.1 | - | 4 | 1039 | |||
M76675.1 | - | 4 | 829 | |||
M81797.1 | - | 4 | 873 | |||
M84425.1 | - | 4 | 951 | |||
M84426.1 | - | 4 | 951 | |||
S62045.1 | - | 4 | 893 |
Set Membership: |
![]() |