Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133296.1 | NP_579830.1 | 7 | 1088 | ||
GenBank mRNA | FQ211786.1 | - | 1196 | |||
S76742.1 | - | 7 | 1088 |
Gene Symbol: RGD1566368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729986.4 | XP_002730032.2 | 7 | 1971 | ||
XM_006244213.2 | XP_006244275.1 | 7 | 1591 | |||
XM_008766730.1 | XP_008764952.1 | 8 | 1482 | |||
XM_008766731.1 | XP_008764953.1 | 7 | 1705 |