Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hmgn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007020.2 | NP_001007021.1 | 4, 5 | 273 | ||
NM_001201353.1 | NP_001188282.1 | 4, 5 | 273 | |||
NM_001201354.1 | NP_001188283.1 | 4, 5 | 273 | |||
NM_001201355.1 | NP_001188284.1 | 4, 5 | 273 | |||
XM_006243445.2 | XP_006243507.1 | 4, 5 | 778 | |||
XM_006243446.2 | XP_006243508.1 | 4, 5 | 778 | |||
XM_008766200.1 | XP_008764422.1 | 4, 5 | 778 | |||
GenBank mRNA | BC082036.1 | - | 4, 5 | 228 | 7130894 | |
CB751211.1 | - | 4, 5 | 156 | |||
CB774488.1 | - | 4, 5 | 241 | |||
FM058030.1 | - | 4, 5 | 273 |