Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190415.1 | NP_001177344.1 | 10 | 1190 | ||
NM_005641.3 | NP_005632.1 | 10 | 1519 | |||
NM_139315.2 | NP_647476.1 | 10 | 1221 | |||
NR_033792.1 | 10 | 1070 | ||||
XM_006716100.2 | XP_006716163.1 | 11 | 1312 | |||
XM_006716101.2 | XP_006716164.1 | 10 | 1149 | |||
XM_006716102.2 | XP_006716165.1 | 10 | 1016 | |||
XM_011516542.2 | XP_011514844.1 | 11 | 1291 | |||
XM_011516543.2 | XP_011514845.1 | 10 | 1044 | |||
XM_017012568.1 | XP_016868057.1 | 10 | 1489 | |||
GenBank mRNA | AK097781.1 | - | 10 | 1072 | ||
AK300005.1 | - | 10 | 1173 | |||
AK314428.1 | - | 10 | 1496 | |||
AK315077.1 | - | 10 | 1171 | |||
BC018115.1 | - | 10 | 1164 | 3905205 | ||
CR627390.1 | - | 10 | 1048 | |||
L25444.1 | - | 10 | 1491 | |||
U31659.1 | - | 9 | 1275 |