Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TAF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006284.3 | NP_006275.1 | 4, 5 | 630 | ||
GenBank mRNA | AL833496.1 | - | 2, 3 | 685 | ||
BC012088.1 | - | 4, 5 | 587 | 4562773 | ||
CR541943.1 | - | 4, 5 | 575 | |||
U13991.1 | - | 4, 5 | 592 |