Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232237.2 | XP_006232299.1 | 2 | 1658 | ||
XM_008760948.1 | XP_008759170.1 | 1 | 2664 | |||
XM_008760949.1 | XP_008759171.1 | 1 | 2662 |