Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtnr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053676.2 | NP_446128.1 | 2 | 581 | ||
GenBank mRNA | AB377274.1 | - | 2 | 582 | ||
AF130341.1 | - | 2 | 357 |
Set Membership: |
![]() |