Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270488.1 | NP_001257417.1 | 2 | 336 | ||
NM_005643.3 | NP_005634.1 | 2 | 336 | |||
XM_011514827.2 | XP_011513129.1 | 2 | 726 | |||
GenBank mRNA | AF118094.1 | - | 2 | 76 | ||
AK302226.1 | - | 2 | 307 | |||
AK313475.1 | - | 2 | 277 | |||
BC021972.2 | - | 2 | 292 | 3935882 | ||
BT019404.1 | - | 2 | 222 | |||
BT019405.1 | - | 2 | 222 | |||
BX538195.1 | - | 1 | 133 | |||
BX647568.1 | - | 1 | 504 | |||
CR542068.1 | - | 2 | 222 | |||
D63705.1 | - | 2 | 305 | |||
DA153455.1 | - | 2 | 336 | |||
X83928.1 | - | 2 | 314 |