Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cdkn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080782.3 | NP_542960.1 | 2, 3 | 486 | ||
XM_006256128.2 | XP_006256190.1 | 2, 3 | 550 | |||
GenBank mRNA | BC100620.1 | - | 2, 3 | 486 | 7938247 | |
U24174.1 | - | 2, 3 | 529 |