Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053769.3 | NP_446221.2 | 3 | 794 | ||
GenBank mRNA | S81478.1 | - | 3 | 768 | ||
U02553.1 | - | 3 | 780 | |||
X84004.1 | - | 3 | 817 |