Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133383.1 | NP_596874.1 | 3 | 272 | ||
GenBank mRNA | AF330051.1 | - | 3 | 272 |