Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053791.2 | NP_446243.1 | 2 | 141 | ||
GenBank mRNA | AF226608.1 | - | 2 | 139 | ||
BC097296.1 | - | 2 | 141 | 7445776 | ||
DQ329279.1 | - | 2 | 139 |