Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053819.1 | NP_446271.1 | 3, 4 | 228 | ||
XM_006256608.2 | XP_006256670.1 | 3, 4 | 292 | |||
XM_008773034.1 | XP_008771256.1 | 2, 3 | 202 | |||
GenBank mRNA | AY550026.1 | - | 2, 3 | 217 | ||
BC099821.1 | - | 3, 4 | 226 | 7930800 | ||
FQ221763.1 | - | 3, 4 | 238 | |||
FQ222271.1 | - | 3, 4 | 238 | |||
FQ222437.1 | - | 3, 4 | 241 | |||
FQ222915.1 | - | 3, 4 | 238 | |||
FQ228400.1 | - | 3, 4 | 237 | |||
FQ230039.1 | - | 3, 4 | 235 | |||
L29512.1 | - | 2, 3 | 106 | |||
L31883.1 | - | 3, 4 | 228 | |||
U06179.1 | - | 2, 3 | 211 | |||
U16022.1 | - | 1, 2 | 122 |