Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phox2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053869.1 | NP_446321.1 | 1, 2 | 410 | ||
GenBank mRNA | U25967.1 | - | 1, 2 | 410 |