Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kcnj4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053870.2 | NP_446322.2 | 2 | 943 | ||
XM_006241963.2 | XP_006242025.1 | 1 | 388 | |||
XM_006241964.2 | XP_006242026.1 | 2 | 1668 | |||
GenBank mRNA | X83580.1 | - | 1 | 345 | ||
X87635.1 | - | 2 | 943 |