Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053874.1 | NP_446326.1 | 12 | 1288 | ||
XM_006253753.2 | XP_006253815.1 | 13 | 1397 | |||
XM_006253754.2 | XP_006253816.1 | 13 | 1510 | |||
GenBank mRNA | U31935.1 | - | 12 | 1288 |