Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053887.1 | NP_446339.1 | 4 | 1485 | ||
GenBank mRNA | JQ013733.1 | - | 4 | 1002 | ||
U48596.1 | - | 4 | 1485 |
Gene Symbol: LOC100912399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231926.2 | XP_006231988.2 | 1466 |