Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053912.2 | NP_446364.2 | 1 | 225 | ||
GenBank mRNA | BC101884.1 | - | 1 | 225 | 7929513 | |
U83897.1 | - | 1 | 218 |