Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053948.4 | NP_446400.1 | 6 | 528 | ||
XM_006230973.1 | XP_006231035.1 | 8 | 813 | |||
GenBank mRNA | BC060550.1 | - | 6 | 484 | 6890594 | |
BC060595.1 | - | 8 | 822 | 6888231 | ||
FQ223906.1 | - | 8 | 818 | |||
FQ224086.1 | - | 6 | 561 | |||
FQ230390.1 | - | 6 | 491 | |||
FQ232354.1 | - | 6 | 532 | |||
Z71925.1 | - | 6 | 529 |