Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053960.3 | NP_446412.2 | 2 | 1087 | ||
GenBank mRNA | BC078756.1 | - | 1 | 1022 | 7109679 | |
Y12009.1 | - | 2 | 1097 |
Set Membership: |
![]() |