Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191915.1 | NP_001178844.1 | 2 | 656 | ||
GenBank mRNA | U52218.1 | - | 1 | 229 |
Set Membership: |
![]() |