Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF4 SIRNA Assays ›
Gene Symbol:
TCF4
Gene Name:
transcription factor 4
Gene Aliases:
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location:
Chr.18: 55222331 - 55635993 on Build GRCh38
UniGene ID:
Hs.605153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s13863
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083962.1
NP_001077431.1
17
2149
NM_001243226.2
NP_001230155.2
18
1939
NM_001243227.1
NP_001230156.1
16
1983
NM_001243228.1
NP_001230157.1
17
2093
NM_001243230.1
NP_001230159.1
16
1622
NM_001243231.1
NP_001230160.1
15
1583
NM_001243232.1
NP_001230161.1
13
1605
NM_001243233.1
NP_001230162.1
13
1546
NM_001243234.1
NP_001230163.1
10
1178
NM_001243235.1
NP_001230164.1
10
1178
NM_001243236.1
NP_001230165.1
10
1194
NM_001306207.1
NP_001293136.1
16
1774
NM_001306208.1
NP_001293137.1
13
1605
NM_003199.2
NP_003190.1
17
2149
XM_005266739.3
XP_005266796.2
16
1789
XM_005266741.3
XP_005266798.1
17
1749
XM_005266743.4
XP_005266800.1
16
1610
XM_005266744.3
XP_005266801.1
16
1730
XM_005266745.3
XP_005266802.1
16
1592
XM_005266747.3
XP_005266804.1
15
1586
XM_005266749.3
XP_005266806.1
13
1377
XM_005266752.4
XP_005266809.1
13
1947
XM_005266754.3
XP_005266811.1
12
1371
XM_005266755.4
XP_005266812.1
13
1406
XM_005266761.3
XP_005266818.1
10
1687
XM_006722536.2
XP_006722599.1
17
2072
XM_006722537.2
XP_006722600.1
17
1705
XM_006722538.2
XP_006722601.1
16
1727
XM_011526158.2
XP_011524460.1
16
2703
XM_011526160.2
XP_011524462.1
12
1756
XM_017025934.1
XP_016881423.1
16
1786
XM_017025935.1
XP_016881424.1
16
1786
XM_017025936.1
XP_016881425.1
16
1789
XM_017025937.1
XP_016881426.1
16
1543
XM_017025938.1
XP_016881427.1
17
1702
XM_017025939.1
XP_016881428.1
17
1749
XM_017025940.1
XP_016881429.1
17
2072
XM_017025941.1
XP_016881430.1
17
1705
XM_017025942.1
XP_016881431.1
17
1749
XM_017025943.1
XP_016881432.1
17
1749
XM_017025944.1
XP_016881433.1
16
2007
XM_017025945.1
XP_016881434.1
16
2007
XM_017025946.1
XP_016881435.1
16
2024
XM_017025947.1
XP_016881436.1
16
1610
XM_017025948.1
XP_016881437.1
16
2007
XM_017025949.1
XP_016881438.1
16
1727
XM_017025950.1
XP_016881439.1
16
2004
XM_017025951.1
XP_016881440.1
15
1583
XM_017025952.1
XP_016881441.1
13
1597
XM_017025953.1
XP_016881442.1
13
1377
XM_017025954.1
XP_016881443.1
13
1958
XM_017025955.1
XP_016881444.1
12
1692
XM_017025956.1
XP_016881445.1
12
1371
XM_017025957.1
XP_016881446.1
10
1687
GenBank mRNA
AB209741.1
-
12
1257
AK095041.1
-
13
1605
AK096862.1
-
15
1583
AK122765.1
-
17
2149
AK299169.1
-
16
1983
AK300038.1
-
10
1124
AK300612.1
-
13
1546
AK300636.1
-
12
1276
AK301144.1
-
16
1760
AK315074.1
-
18
2081
AK316165.1
-
12
1320
BC031056.1
-
18
1900
BC125084.1
-
16
1563
40117795
BC125085.1
-
16
1563
40117798
FR748210.1
-
16
1537
FR748211.1
-
16
1537
FR748212.1
-
14
1357
FR748213.1
-
14
1357
FR748214.1
-
15
1465
FR748215.1
-
13
1285
FR748216.1
-
16
1528
FR748217.1
-
15
1411
FR748218.1
-
11
1147
FR748219.1
-
13
1324
FR748220.1
-
10
1057
FR748221.1
-
10
1057
FR748222.1
-
10
1057
FR748223.1
-
8
889
M74718.1
-
10
1009
M74719.1
-
17
1736
X52079.1
-
14
1402
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, TFIIB-class binding
TFIIB-class transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein C-terminus binding
identical protein binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-vrvhj:80/10.239.161.228:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0