Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF4 SIRNA Assays ›
Gene Symbol:
TCF4
Gene Name:
transcription factor 4
Gene Aliases:
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location:
Chr.18: 55222331 - 55635993 on Build GRCh38
UniGene ID:
Hs.605153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s13864
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083962.1
NP_001077431.1
14
1746
NM_001243226.2
NP_001230155.2
15
1536
NM_001243227.1
NP_001230156.1
13
1580
NM_001243228.1
NP_001230157.1
14
1690
NM_001243230.1
NP_001230159.1
13
1219
NM_001243231.1
NP_001230160.1
12
1180
NM_001243232.1
NP_001230161.1
10
1202
NM_001243233.1
NP_001230162.1
10
1143
NM_001243234.1
NP_001230163.1
7
775
NM_001243235.1
NP_001230164.1
7
775
NM_001243236.1
NP_001230165.1
7
791
NM_001306207.1
NP_001293136.1
13
1371
NM_001306208.1
NP_001293137.1
10
1202
NM_003199.2
NP_003190.1
14
1746
XM_005266739.3
XP_005266796.2
13
1386
XM_005266741.3
XP_005266798.1
14
1346
XM_005266743.4
XP_005266800.1
13
1207
XM_005266744.3
XP_005266801.1
13
1327
XM_005266745.3
XP_005266802.1
13
1189
XM_005266747.3
XP_005266804.1
12
1183
XM_005266749.3
XP_005266806.1
10
974
XM_005266752.4
XP_005266809.1
10
1544
XM_005266754.3
XP_005266811.1
9
968
XM_005266755.4
XP_005266812.1
10
1003
XM_005266761.3
XP_005266818.1
7
1284
XM_006722536.2
XP_006722599.1
14
1669
XM_006722537.2
XP_006722600.1
14
1302
XM_006722538.2
XP_006722601.1
13
1324
XM_011526158.2
XP_011524460.1
13
2300
XM_011526160.2
XP_011524462.1
9
1353
XM_017025934.1
XP_016881423.1
13
1383
XM_017025935.1
XP_016881424.1
13
1383
XM_017025936.1
XP_016881425.1
13
1386
XM_017025937.1
XP_016881426.1
13
1140
XM_017025938.1
XP_016881427.1
14
1299
XM_017025939.1
XP_016881428.1
14
1346
XM_017025940.1
XP_016881429.1
14
1669
XM_017025941.1
XP_016881430.1
14
1302
XM_017025942.1
XP_016881431.1
14
1346
XM_017025943.1
XP_016881432.1
14
1346
XM_017025944.1
XP_016881433.1
13
1604
XM_017025945.1
XP_016881434.1
13
1604
XM_017025946.1
XP_016881435.1
13
1621
XM_017025947.1
XP_016881436.1
13
1207
XM_017025948.1
XP_016881437.1
13
1604
XM_017025949.1
XP_016881438.1
13
1324
XM_017025950.1
XP_016881439.1
13
1601
XM_017025951.1
XP_016881440.1
12
1180
XM_017025952.1
XP_016881441.1
10
1194
XM_017025953.1
XP_016881442.1
10
974
XM_017025954.1
XP_016881443.1
10
1555
XM_017025955.1
XP_016881444.1
9
1289
XM_017025956.1
XP_016881445.1
9
968
XM_017025957.1
XP_016881446.1
7
1284
GenBank mRNA
AB209741.1
-
9
854
AK095041.1
-
10
1202
AK096862.1
-
12
1180
AK122765.1
-
14
1746
AK299169.1
-
13
1580
AK300038.1
-
7
721
AK300612.1
-
10
1143
AK300636.1
-
9
873
AK301144.1
-
13
1357
AK315074.1
-
15
1678
AK316165.1
-
9
917
AL518903.3
-
7
791
BC031056.1
-
15
1497
BC125084.1
-
13
1160
40117795
BC125085.1
-
13
1160
40117798
FR748210.1
-
13
1134
FR748211.1
-
13
1134
FR748212.1
-
11
954
FR748213.1
-
11
954
FR748214.1
-
12
1062
FR748215.1
-
10
882
FR748216.1
-
13
1125
FR748217.1
-
12
1008
FR748218.1
-
8
744
FR748219.1
-
10
921
FR748220.1
-
7
654
FR748221.1
-
7
654
FR748222.1
-
7
654
FR748223.1
-
5
486
M74718.1
-
7
606
M74719.1
-
14
1333
X52079.1
-
11
999
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, TFIIB-class binding
TFIIB-class transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein C-terminus binding
identical protein binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-tm9kw:80/10.239.162.147:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0