Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tra2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057119.1 | NP_476460.1 | 4 | 622 | ||
XM_006248536.2 | XP_006248598.1 | 4 | 663 | |||
XM_008768789.1 | XP_008767011.1 | 4 | 4778 | |||
XM_008768790.1 | XP_008767012.1 | 5 | 939 | |||
GenBank mRNA | BC070948.1 | - | 4 | 622 | 7097246 | |
D49708.1 | - | 4 | 622 |