Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057137.1 | NP_476478.1 | 3, 4 | 436 | ||
XM_006256699.2 | XP_006256761.1 | 3, 4 | 461 | |||
GenBank mRNA | AF071501.1 | - | 3, 4 | 436 | ||
FQ209542.1 | - | 1, 2 | 18 | |||
FQ219951.1 | - | 1, 2 | 18 | |||
FQ230059.1 | - | 1, 2 | 18 |