Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Park7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277249.1 | NP_001264178.1 | 6 | 475 | ||
NM_001277250.1 | NP_001264179.1 | 6 | 475 | |||
NM_001277251.1 | NP_001264180.1 | 7 | 534 | |||
NM_001277252.1 | NP_001264181.1 | 6 | 415 | |||
NM_001277253.1 | NP_001264182.1 | 6 | 527 | |||
NM_057143.2 | NP_476484.1 | 6 | 657 | |||
GenBank mRNA | AF157511.1 | - | 6 | 657 | ||
AF157512.1 | - | 6 | 411 | |||
AJ007291.1 | - | 7 | 582 | |||
BC091128.1 | - | 6 | 448 | 7376914 | ||
FM055257.1 | - | 6 | 445 | |||
FM061417.1 | - | 6 | 396 | |||
FM069664.1 | - | 6 | 466 | |||
FQ217993.1 | - | 2 | 104 | |||
FQ220895.1 | - | 6 | 527 | |||
FQ223564.1 | - | 6 | 525 | |||
FQ224480.1 | - | 7 | 518 |