Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF7 SIRNA Assays ›
Gene Symbol:
TCF7
Gene Name:
transcription factor 7 (T-cell specific, HMG-box)
Gene Aliases:
TCF-1
Chromosome Location:
Chr.5: 134114703 - 134148229 on Build GRCh38
UniGene ID:
Hs.573153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s13877
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134851.2
NP_001128323.2
5, 6
504
NM_003202.3
NP_003193.2
6, 7
948
NM_201632.3
NP_963963.1
5, 6
504
NM_201634.3
NP_963965.1
5, 6
504
NM_213648.3
NP_998813.1
4, 5
738
NR_033449.1
4, 5
739
XM_006714678.3
XP_006714741.1
7, 8
1049
XM_006714679.3
XP_006714742.1
6, 7
956
XM_006714680.3
XP_006714743.1
7, 8
1049
XM_006714682.2
XP_006714745.1
7, 8
1049
XM_006714684.2
XP_006714747.1
6, 7
956
XM_006714685.3
XP_006714748.1
6, 7
594
XM_006714686.3
XP_006714749.1
5, 6
503
XM_006714687.3
XP_006714750.1
5, 6
461
XM_011543604.2
XP_011541906.1
7, 8
1049
XM_011543606.1
XP_011541908.1
7, 8
1049
XM_011543607.2
XP_011541909.1
5, 6
6537
XM_011543608.2
XP_011541910.1
5, 6
2582
XM_011543609.2
XP_011541911.1
4, 5
450
XM_011543610.2
XP_011541912.1
5, 6
582
XM_011543611.2
XP_011541913.1
5, 6
1476
XM_011543612.2
XP_011541914.1
4, 5
450
XM_011543613.2
XP_011541915.1
4, 5
489
XM_017009789.1
XP_016865278.1
6, 7
594
XM_017009790.1
XP_016865279.1
5, 6
1476
GenBank mRNA
AK057580.1
-
4, 5
739
AK093530.1
-
4, 5
609
AK093683.1
-
1, 2
838
AK131428.1
-
2, 3
471
AK310591.1
-
4, 5
450
AL832816.1
-
6, 7
664
AL834166.1
-
6, 7
565
BC048769.1
-
6, 7
948
6063270
BC072023.1
-
6, 7
883
6292932
DA627229.1
-
5, 6
504
DB145426.1
-
5, 6
504
X59869.1
-
5, 6
486
X59870.1
-
5, 6
486
X59871.1
-
5, 6
486
Z47361.1
-
4, 5
408
Z47362.1
-
4, 5
408
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
neural tube development
embryonic genitalia morphogenesis
T cell receptor V(D)J recombination
regulation of cell proliferation
canonical Wnt signaling pathway involved in negative regulation of apoptotic process
alpha-beta T cell differentiation
embryonic digestive tract morphogenesis
embryonic hindgut morphogenesis
canonical Wnt signaling pathway
cellular response to interleukin-4
beta-catenin-TCF complex assembly
core promoter binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein binding
beta-catenin binding
sequence-specific DNA binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0