Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF7L2 SIRNA Assays ›
Gene Symbol:
TCF7L2
Gene Name:
transcription factor 7 like 2
Gene Aliases:
TCF-4, TCF4
Chromosome Location:
Chr.10: 112950219 - 113167678 on Build GRCh38
UniGene ID:
Hs.593995
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s13880
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF7L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146274.1
NP_001139746.1
14
1971
NM_001146283.1
NP_001139755.1
13
1970
NM_001146284.1
NP_001139756.1
13
1868
NM_001146285.1
NP_001139757.1
13
1902
NM_001146286.1
NP_001139758.1
12
1829
NM_001198525.1
NP_001185454.1
15
2041
NM_001198526.1
NP_001185455.1
13
1902
NM_001198527.1
NP_001185456.1
15
1966
NM_001198528.1
NP_001185457.1
15
1978
NM_001198529.1
NP_001185458.1
13
1880
NM_001198530.1
NP_001185459.1
11
1727
NM_001198531.1
NP_001185460.1
13
1898
NM_030756.4
NP_110383.2
14
1953
XM_005270071.1
XP_005270128.1
16
2212
XM_005270075.1
XP_005270132.1
16
2197
XM_005270077.1
XP_005270134.1
15
2161
XM_005270078.1
XP_005270135.1
15
2161
XM_005270079.1
XP_005270136.1
15
2146
XM_005270080.1
XP_005270137.1
15
2143
XM_005270084.1
XP_005270141.1
15
2053
XM_005270085.1
XP_005270142.1
14
2017
XM_005270086.1
XP_005270143.1
14
1999
XM_005270088.1
XP_005270145.1
14
1972
XM_005270089.1
XP_005270146.1
13
1948
XM_005270091.2
XP_005270148.1
17
2285
XM_005270093.2
XP_005270150.1
15
2139
XM_005270094.2
XP_005270151.1
15
2136
XM_005270095.1
XP_005270152.1
14
2088
XM_005270096.2
XP_005270153.1
14
1980
XM_005270100.1
XP_005270157.1
13
1944
XM_005270101.2
XP_005270158.1
13
1926
XM_005270102.1
XP_005270159.1
13
1917
XM_005270103.1
XP_005270160.1
12
1875
XM_005270104.1
XP_005270161.1
12
1848
XM_011540109.1
XP_011538411.1
16
2212
XM_011540110.1
XP_011538412.1
15
2116
XM_011540111.1
XP_011538413.1
14
1984
XM_011540116.1
XP_011538418.1
16
2234
XM_017016584.1
XP_016872073.1
16
2209
XM_017016585.1
XP_016872074.1
16
2200
XM_017016586.1
XP_016872075.1
16
2197
XM_017016587.1
XP_016872076.1
16
2185
XM_017016588.1
XP_016872077.1
15
2143
XM_017016589.1
XP_016872078.1
14
2089
XM_017016590.1
XP_016872079.1
15
2068
XM_017016591.1
XP_016872080.1
15
2056
XM_017016592.1
XP_016872081.1
15
2053
XM_017016593.1
XP_016872082.1
14
2002
XM_017016594.1
XP_016872083.1
14
1999
XM_017016595.1
XP_016872084.1
14
2073
XM_017016596.1
XP_016872085.1
14
1968
GenBank mRNA
AB440195.1
-
13
1391
AB451266.1
-
14
1446
AK074705.1
-
4
344
AK225809.1
-
13
1906
AK296305.1
-
12
1343
AK299295.1
-
12
1359
AK301483.1
-
11
1169
AK315243.1
-
13
1636
AK316111.1
-
12
1056
BC032656.1
-
13
1457
5533185
BX648364.1
-
14
1339
CR536574.1
-
14
1446
FJ010164.1
-
13
1341
FJ010165.1
-
14
1473
FJ010166.1
-
15
1627
FJ010167.1
-
13
1488
FJ010170.1
-
13
1466
FJ010171.1
-
11
1257
FJ010172.1
-
12
1419
FJ010173.1
-
13
1341
FJ010174.1
-
14
1850
HM352838.1
-
13
1388
HM352839.1
-
13
1410
HM352840.1
-
13
1379
HM352841.1
-
13
1406
HM352842.1
-
12
1322
HM352843.1
-
12
1310
HM352844.1
-
14
1442
HM352845.1
-
15
1575
HM352846.1
-
13
1391
HM352847.1
-
13
1395
HM352848.1
-
14
1446
HM352849.1
-
12
1337
HM352850.1
-
15
1515
HM352851.1
-
14
1479
Y11306.2
-
14
1753
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
blood vessel development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell cycle arrest
Wnt signaling pathway, calcium modulating pathway
cell proliferation
response to glucose
positive regulation of heparan sulfate proteoglycan biosynthetic process
pancreas development
positive regulation of insulin secretion
positive regulation of protein binding
regulation of hormone metabolic process
glucose homeostasis
negative regulation of sequence-specific DNA binding transcription factor activity
maintenance of DNA repeat elements
canonical Wnt signaling pathway involved in positive regulation of epithelial to mesenchymal transition
fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein export from nucleus
myoblast fate commitment
regulation of smooth muscle cell proliferation
positive regulation of protein kinase B signaling
canonical Wnt signaling pathway
negative regulation of canonical Wnt signaling pathway
beta-catenin-TCF complex assembly
negative regulation of type B pancreatic cell apoptotic process
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II repressing transcription factor binding
transcription factor activity, sequence-specific DNA binding
protein binding
beta-catenin binding
transcription factor binding
protein kinase binding
nuclear hormone receptor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
gamma-catenin binding
armadillo repeat domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0