Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057200.2 | NP_476548.1 | 16 | 1808 | ||
XM_008764204.1 | XP_008762426.1 | 16 | 1814 | |||
XM_008764205.1 | XP_008762427.1 | 14 | 1518 | |||
GenBank mRNA | AB070355.1 | - | 16 | 1808 | ||
AB120429.1 | - | 13 | 1316 | |||
AF083331.1 | - | 13 | 1397 |