Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacng3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080691.1 | NP_542422.1 | 1 | 130 | ||
GenBank mRNA | AF361340.1 | - | 1 | 130 |