Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001062.3 | NP_001053.2 | 7 | 1091 | ||
GenBank mRNA | AK292990.1 | - | 7 | 1008 | ||
BC018632.1 | - | 7 | 1013 | |||
BM769693.1 | - | 3 | 337 | |||
J05068.1 | - | 7 | 1068 |