Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130410.2 | NP_569094.1 | 1, 2 | 469 | ||
GenBank mRNA | AY055379.1 | - | 1, 2 | 197 | ||
BC098907.1 | - | 1, 2 | 469 | 7454618 |