Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130411.2 | NP_569095.1 | 8 | 981 | ||
XM_006230204.2 | XP_006230266.1 | 8 | 1065 | |||
GenBank mRNA | AF416730.1 | - | 7 | 877 | ||
AF495469.1 | - | 8 | 981 | |||
BC086971.1 | - | 8 | 957 | 7124429 | ||
FQ225277.1 | - | 5 | 503 | |||
FQ225360.1 | - | 5 | 503 | |||
FQ226290.1 | - | 6 | 450 | |||
FQ226379.1 | - | 8 | 972 | |||
FQ226941.1 | - | 8 | 1001 | |||
FQ233306.1 | - | 8 | 975 | |||
FQ233390.1 | - | 8 | 977 | |||
FQ233980.1 | - | 8 | 895 | |||
FQ234750.1 | - | 8 | 983 |