Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130420.1 | NP_569104.1 | 8 | 2016 | ||
XM_008764691.1 | XP_008762913.1 | 12 | 2345 | |||
XM_008764694.1 | XP_008762916.1 | 8 | 2045 | |||
GenBank mRNA | AF350422.1 | - | 8 | 2016 |