Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmprss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130424.2 | NP_569108.2 | 8 | 805 | ||
XM_008768547.1 | XP_008766769.1 | 8 | 921 | |||
XM_008768548.1 | XP_008766770.1 | 8 | 852 | |||
XM_008768549.1 | XP_008766771.1 | 8 | 1034 | |||
GenBank mRNA | AB073550.1 | - | 6 | 703 | ||
BC061712.1 | - | 8 | 805 | 5598010 |