Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Eya2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130427.1 | NP_569111.1 | 14 | 1451 | ||
XM_008762456.1 | XP_008760678.1 | 15 | 1687 | |||
GenBank mRNA | AY366465.1 | - | 14 | 1451 |