Search Thermo Fisher Scientific
Gene Symbol: Hrh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131909.1 | NP_571984.1 | 1 | 178 | ||
GenBank mRNA | AF358860.1 | - | 1 | 178 |
Set Membership: |
G-protein coupled receptor |