Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynlrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131910.3 | NP_571985.1 | 3 | 334 | ||
GenBank mRNA | AF073839.2 | - | 3 | 257 | ||
AY026512.1 | - | 3 | 334 | |||
BC058437.1 | - | 3 | 275 | 6921076 |