Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mutyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133316.1 | NP_579850.1 | 5 | 410 | ||
XM_006238626.2 | XP_006238688.1 | 5 | 438 | |||
XM_006238627.2 | XP_006238689.1 | 5 | 449 | |||
XM_008763987.1 | XP_008762209.1 | 4 | 295 | |||
GenBank mRNA | AF478683.1 | - | 5 | 410 | ||
BC128728.1 | - | 5 | 340 | |||
FQ230684.1 | - | 4 | 517 |