Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133390.1 | NP_596881.1 | 5 | 3333 | ||
GenBank mRNA | AB047637.1 | - | 5 | 3333 |