Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133398.1 | NP_596889.1 | 4 | 1016 | ||
XM_006241520.2 | XP_006241582.1 | 4 | 2435 | |||
XM_006241521.2 | XP_006241583.1 | 4 | 2436 | |||
XM_006241523.2 | XP_006241585.1 | 4 | 2433 | |||
XM_006241524.2 | XP_006241586.1 | 4 | 2433 | |||
XM_006241525.2 | XP_006241587.1 | 4 | 2434 | |||
GenBank mRNA | AF100421.2 | - | 4 | 1016 | ||
FQ225234.1 | - | 2 | 164 | |||
FQ225997.1 | - | 2 | 164 | |||
FQ226094.1 | - | 4 | 1012 | |||
FQ227915.1 | - | 2 | 164 | |||
FQ231703.1 | - | 2 | 164 | |||
FQ234551.1 | - | 4 | 1020 |