Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133418.1 | NP_596909.1 | 6 | 473 | ||
GenBank mRNA | AJ223355.1 | - | 6 | 473 | ||
BC081734.1 | - | 6 | 491 | 7108552 |