Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005651.3 | NP_005642.1 | 6 | 602 | ||
GenBank mRNA | AK289418.1 | - | 6 | 599 | ||
AK307741.1 | - | 6 | 599 | |||
BC005355.1 | - | 6 | 599 | 4071714 | ||
U32989.1 | - | 6 | 600 |